Learn More
Biomatik Corporation Rat Pentraxin 3, Long (PTX3) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06504
Encompass_Preferred
Rat Pentraxin 3, Long (PTX3) ELISA Kit for detecting PTX3 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 31.2-2,000pg/ml. Sensitivity: 13.6pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: D3ZT94. Gene ID: 689388. Target Synonyms: TNFAIP5; TSG-14; Pentaxin-Related Gene,Rapidly Induced By IL-1 Beta; Tumor Necrosis Factor Alpha-Induced Protein 5; Tumor necrosis factor-inducible gene 14 protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3469