Learn More
Biomatik Corporation Rat Pepsinogen A (PGA) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06508
Encompass_Preferred
Rat Pepsinogen A (PGA) ELISA Kit for detecting PGA in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.62ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9JJX2. Gene ID: 60372. Target Synonyms: PGA5; PG-A; Pepsinogen 5 Group I; Pepsinogen-5; Pepsin A-5. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3470