Learn More
Biomatik Corporation Human Peptidase Inhibitor 3, Skin Derived (PI3) ELISA Kit, 96 tests
Catalog No. 501503471
Encompass_Preferred
The Human Peptidase Inhibitor 3, Skin Derived (PI3) ELISA Kit allows for quantitative determination of Peptidase Inhibitor 3, Skin Derived in Serum, Plasma, Urine, Saliva, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.124 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P19957. Cross Activity: No significant cross-reactivity or interference between Peptidase Inhibitor 3, Skin Derived and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3471 Supplier Biomatik Corporation Supplier No. EKU06514
The Human Peptidase Inhibitor 3, Skin Derived (PI3) ELISA Kit allows for quantitative determination of Peptidase Inhibitor 3, Skin Derived in Serum, Plasma, Urine, Saliva, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.124 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P19957. Cross Activity: No significant cross-reactivity or interference between Peptidase Inhibitor 3, Skin Derived and analogues was observed. Restriction: For Research Use Only.