Learn More
Biomatik Corporation Human Peptide YY (PYY) ELISA Kit, 96 tests
Catalog No. 501503472
Encompass_Preferred
The Human Peptide YY (PYY) ELISA Kit allows for quantitative determination of Peptide YY in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 12.35-1,000 pg/mL. Sensitivity: 4.77 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. UniProt ID: P10082. Cross Activity: No significant cross-reactivity or interference between Peptide YY and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3472 Supplier Biomatik Corporation Supplier No. EKU06515
The Human Peptide YY (PYY) ELISA Kit allows for quantitative determination of Peptide YY in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 12.35-1,000 pg/mL. Sensitivity: 4.77 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. UniProt ID: P10082. Cross Activity: No significant cross-reactivity or interference between Peptide YY and analogues was observed. Restriction: For Research Use Only.