Learn More
Biomatik Corporation Rat Peptide YY (PYY) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06517
Encompass_Preferred
Rat Peptide YY (PYY) ELISA Kit for detecting PYY in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 12.35-1,000pg/ml. Sensitivity: 4.94pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P10631. Gene ID: 287730. Target Synonyms: PYY1; Peptide tyrosine tyrosine. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3473