Learn More
Biomatik Corporation Human Peptidoglycan Recognition Protein 1 (PGLYRP1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06519
Encompass_Preferred
The Human Peptidoglycan Recognition Protein 1 (PGLYRP1) ELISA Kit allows for quantitative determination of Peptidoglycan Recognition Protein 1 in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.78-50 ng/mL. Sensitivity: 0.31 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: O75594. Cross Activity: No significant cross-reactivity or interference between Peptidoglycan Recognition Protein 1 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3474