Learn More
Biomatik Corporation Mouse Peptidoglycan Recognition Protein 1 (PGLYRP1) ELISA Kit, 96 tests
Catalog No. 501503475
Encompass_Preferred
Mouse Peptidoglycan Recognition Protein 1 (PGLYRP1) ELISA Kit for detecting PGLYRP1 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.7pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O88593. Gene ID: 21946. Target Synonyms: PGLYRP; PGRP; PGRP-S; PGRPS; TAG7; TNFSF3L; Tumor Necrosis Factor Ligand Superfamily Member 3 Like; Peptidoglycan recognition protein short. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3475 Supplier Biomatik Corporation Supplier No. EKU06520
Mouse Peptidoglycan Recognition Protein 1 (PGLYRP1) ELISA Kit for detecting PGLYRP1 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.7pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O88593. Gene ID: 21946. Target Synonyms: PGLYRP; PGRP; PGRP-S; PGRPS; TAG7; TNFSF3L; Tumor Necrosis Factor Ligand Superfamily Member 3 Like; Peptidoglycan recognition protein short. Restrictions: For Research Use Only. Not for use in diagnostic procedures.