Learn More
Biomatik Corporation ABL1 Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50011
Encompass_Preferred
ABL1 Colorimetric Cell-Based ELISA Kit for detecting ABL1 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P00519. Gene ID: 25. Target Synonyms: ABL1; JTK7; Proto-oncogene tyrosine-protein kinase ABL1; c-ABL; p150. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8892