Learn More
Biomatik Corporation Human Peptidoglycan Recognition Protein 2 (PGLYRP2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06521
Encompass_Preferred
The Human Peptidoglycan Recognition Protein 2 (PGLYRP2) ELISA Kit allows for quantitative determination of Peptidoglycan Recognition Protein 2 in Serum, Plasma, Urine, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 15.6-1,000 pg/mL. Sensitivity: 5.7 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q96PD5. Cross Activity: No significant cross-reactivity or interference between Peptidoglycan Recognition Protein 2 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3476