Learn More
Biomatik Corporation AhR Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50020
Encompass_Preferred
AhR Colorimetric Cell-Based ELISA Kit for detecting AhR in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P35869; A9YTQ3. Gene ID: 196; 57491. Target Synonyms: Aryl hydrocarbon receptor; Ah receptor; AhR; Class E basic helix-loop-helix protein 76; bHLHe76; BHLHE76. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8894