Learn More
Biomatik Corporation Human Peptidylglycine Alpha Amidating Monooxygenase (PAM) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06525
Encompass_Preferred
The Human Peptidylglycine Alpha Amidating Monooxygenase (PAM) ELISA Kit allows for quantitative determination of Peptidylglycine Alpha Amidating Monooxygenase in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 31.2-2,000 pg/mL. Sensitivity: 13.9 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P19021. Cross Activity: No significant cross-reactivity or interference between Peptidylglycine Alpha Amidating Monooxygenase and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3478