Learn More
Biomatik Corporation Claudin 1 Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50121
Encompass_Preferred
Claudin 1 Colorimetric Cell-Based ELISA Kit for detecting Claudin 1 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: O95832. Gene ID: 9076. Target Synonyms: CLD1; CLDN1; SEMP1; Senescence-associated epithelial membrane protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8916