Learn More
Biomatik Corporation Rat Periostin (POSTN) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06540
Encompass_Preferred
Rat Periostin (POSTN) ELISA Kit for detecting POSTN in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 31.2-2,000pg/ml. Sensitivity: 13.1pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: D3ZAF5. Gene ID: 361945. Target Synonyms: OSF-2; PDLPOSTN; PN; Osteoblast Specific Factor 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3485