Learn More
Biomatik Corporation Collagen I α2 Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50133
Encompass_Preferred
Collagen I α2 Colorimetric Cell-Based ELISA Kit for detecting Collagen I α2 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P08123. Gene ID: 1278. Target Synonyms: alpha 2 type i collagen; co1a2; col1a2; collagen type i alpha 2; oi4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8918