Learn More
Biomatik Corporation Human Permeability Glycoprotein (Pgp) ELISA Kit, 96 tests
Catalog No. 501503487
Encompass_Preferred
The Human Permeability Glycoprotein (Pgp) ELISA Kit allows for quantitative determination of Permeability Glycoprotein in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.121 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P08183. Cross Activity: No significant cross-reactivity or interference between Permeability Glycoprotein and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3487 Supplier Biomatik Corporation Supplier No. EKU06547
The Human Permeability Glycoprotein (Pgp) ELISA Kit allows for quantitative determination of Permeability Glycoprotein in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.121 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P08183. Cross Activity: No significant cross-reactivity or interference between Permeability Glycoprotein and analogues was observed. Restriction: For Research Use Only.