Learn More
Biomatik Corporation Human Peroxidasin Homolog (PXDN) ELISA Kit, 96 tests
Catalog No. 501503488
Encompass_Preferred
The Human Peroxidasin Homolog (PXDN) ELISA Kit allows for quantitative determination of Peroxidasin Homolog in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.065 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q92626. Cross Activity: No significant cross-reactivity or interference between Peroxidasin Homolog and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3488 Supplier Biomatik Corporation Supplier No. EKU06549
The Human Peroxidasin Homolog (PXDN) ELISA Kit allows for quantitative determination of Peroxidasin Homolog in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.065 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q92626. Cross Activity: No significant cross-reactivity or interference between Peroxidasin Homolog and analogues was observed. Restriction: For Research Use Only.