Learn More
Biomatik Corporation Rat Peroxiredoxin 1 (PRDX1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06552
Encompass_Preferred
Rat Peroxiredoxin 1 (PRDX1) ELISA Kit for detecting PRDX1 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q63716. Gene ID: 117254. Target Synonyms: MSP23; NKEFA; PAG; PAGA; PAGB; TDPX2; Natural killer cell-enhancing factor A; Proliferation-associated; Thioredoxin peroxidase 2; Thioredoxin-dependent peroxide reductase 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3491