Learn More
Biomatik Corporation Human Peroxiredoxin 2 (PRDX2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06553
Encompass_Preferred
The Human Peroxiredoxin 2 (PRDX2) ELISA Kit allows for quantitative determination of Peroxiredoxin 2 in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 31.2-2000 pg/mL. Sensitivity: 14.2 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P32119. Cross Activity: No significant cross-reactivity or interference between Peroxiredoxin 2 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3492