Learn More
Biomatik Corporation Human Peroxisome Proliferator Activated Receptor Alpha (PPARa) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06559
Encompass_Preferred
The Human Peroxisome Proliferator Activated Receptor Alpha (PPARa) ELISA Kit allows for quantitative determination of Peroxisome Proliferator Activated Receptor Alpha in Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.114 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q07869. Cross Activity: No significant cross-reactivity or interference between Peroxisome Proliferator Activated Receptor Alpha and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3494