Learn More
Biomatik Corporation Human Peroxisome Proliferator Activated Receptor Delta (PPARd) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06562
Encompass_Preferred
The Human Peroxisome Proliferator Activated Receptor Delta (PPARd) ELISA Kit allows for quantitative determination of Peroxisome Proliferator Activated Receptor Delta in Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.115 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q03181. Cross Activity: No significant cross-reactivity or interference between Peroxisome Proliferator Activated Receptor Delta and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3495