Learn More
Biomatik Corporation Rat Peroxisome Proliferator Activated Receptor Gamma (PPARg) ELISA Kit, 96 tests
Catalog No. 501503498
Encompass_Preferred
Rat Peroxisome Proliferator Activated Receptor Gamma (PPARg) ELISA Kit for detecting PPARg in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.054ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O88275. Gene ID: 25664. Target Synonyms: PPAR-G; PPARG1; PPARG2; NR1C3; Glitazone Receptor; Nuclear Receptor Subfamily 1 Group C Member 3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3498 Supplier Biomatik Corporation Supplier No. EKU06567
Rat Peroxisome Proliferator Activated Receptor Gamma (PPARg) ELISA Kit for detecting PPARg in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.054ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O88275. Gene ID: 25664. Target Synonyms: PPAR-G; PPARG1; PPARG2; NR1C3; Glitazone Receptor; Nuclear Receptor Subfamily 1 Group C Member 3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.