Learn More
Biomatik Corporation Rabbit Peroxisome Proliferator Activated Receptor Gamma (PPARg) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06568
Encompass_Preferred
Rabbit Peroxisome Proliferator Activated Receptor Gamma (PPARg) ELISA Kit for detecting PPARg in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rabbit (Oryctolagus cuniculus). Detection Range: 0.78-50ng/ml. Sensitivity: 0.30ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O19052. Target Synonyms: PPAR-G; PPARG1; PPARG2; NR1C3; Glitazone Receptor; Nuclear Receptor Subfamily 1 Group C Member 3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3499