Learn More
Biomatik Corporation Human Peroxisome Proliferator Activated Receptor Gamma Coactivator 1 Alpha (PPARgC1a) ELISA Kit, 96 tests
Catalog No. 501503500
Encompass_Preferred
The Human Peroxisome Proliferator Activated Receptor Gamma Coactivator 1 Alpha (PPARgC1a) ELISA Kit allows for quantitative determination of Peroxisome Proliferator Activated Receptor Gamma Coactivator 1 Alpha in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.055 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q9UBK2. Cross Activity: No significant cross-reactivity or interference between Peroxisome Proliferator Activated Receptor Gamma Coactivator 1 Alpha and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3500 Supplier Biomatik Corporation Supplier No. EKU06569
The Human Peroxisome Proliferator Activated Receptor Gamma Coactivator 1 Alpha (PPARgC1a) ELISA Kit allows for quantitative determination of Peroxisome Proliferator Activated Receptor Gamma Coactivator 1 Alpha in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.055 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q9UBK2. Cross Activity: No significant cross-reactivity or interference between Peroxisome Proliferator Activated Receptor Gamma Coactivator 1 Alpha and analogues was observed. Restriction: For Research Use Only.