Learn More
Biomatik Corporation Mouse Peroxisome Proliferator Activated Receptor Gamma Coactivator 1 Alpha (PPARgC1a) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06570
Encompass_Preferred
Mouse Peroxisome Proliferator Activated Receptor Gamma Coactivator 1 Alpha (PPARgC1a) ELISA Kit for detecting PPARgC1a in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O70343. Target Synonyms: LEM6; PGC-1 (alpha) ; PGC-1v; PGC1; PGC1A; PPARGC1; Ligand effect modulator 6. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3501