Learn More
Biomatik Corporation Mouse Phosphatidylethanolamine Binding Protein 1 (PEBP1) ELISA Kit, 96 tests
Catalog No. 501503507
Encompass_Preferred
Mouse Phosphatidylethanolamine Binding Protein 1 (PEBP1) ELISA Kit for detecting PEBP1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.054ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P70296. Gene ID: 23980. Target Synonyms: PBP; HCNP; PEBP; RKIP; Prostatic Binding Protein; Raf Kinase Inhibitory Protein; Hippocampal Cholinergic Neurostimulating Peptide; Neuropolypeptide h3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3507 Supplier Biomatik Corporation Supplier No. EKU06584
Mouse Phosphatidylethanolamine Binding Protein 1 (PEBP1) ELISA Kit for detecting PEBP1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.054ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P70296. Gene ID: 23980. Target Synonyms: PBP; HCNP; PEBP; RKIP; Prostatic Binding Protein; Raf Kinase Inhibitory Protein; Hippocampal Cholinergic Neurostimulating Peptide; Neuropolypeptide h3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.