Learn More
Biomatik Corporation YAP Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50633
Encompass_Preferred
YAP Colorimetric Cell-Based ELISA Kit for detecting YAP in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P46937. Gene ID: 10413. Target Synonyms: 65 kDa Yes-associated protein; YAP1; YAP65. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8982