Learn More
Biomatik Corporation Human Phosphodiesterase 5A, cGMP Specific (PDE5A) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06593
Encompass_Preferred
The Human Phosphodiesterase 5A, cGMP Specific (PDE5A) ELISA Kit allows for quantitative determination of Phosphodiesterase 5A, cGMP Specific in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 1.56-100 ng/mL. Sensitivity: 0.56 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: O76074. Cross Activity: No significant cross-reactivity or interference between Phosphodiesterase 5A, cGMP Specific and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3508