Learn More
Biomatik Corporation Keratin 14 Colorimetric Cell-Based ELISA
Supplier:  Biomatik Corporation EKA50637
Encompass_Preferred
Keratin 14 Colorimetric Cell-Based ELISA for detecting Keratin 14 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P02533; P08779. Gene ID: 3861; 3868; 3872. Target Synonyms: Keratin; type I cytoskeletal 14; Cytokeratin-14; CK-14; K14. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8984