Learn More
Biomatik Corporation Rat Phosphofructokinase, Liver (PFKL) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06598
Encompass_Preferred
Rat Phosphofructokinase, Liver (PFKL) ELISA Kit for detecting PFKL in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.67ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P30835. Gene ID: 25741. Target Synonyms: PFK-B; 6-phosphofructokinase type B; Phosphofructo-1-kinase isozyme B; Phosphohexokinase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3510