Learn More
Biomatik Corporation BDNF (Human) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA52013
Encompass_Preferred
BDNF (Human) Sandwich ELISA Kit for detecting BDNF in plasma, serum, lysate. Species Reactivity: Human (Homo sapiens). Detection Range: 47-1500pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P23560. Gene ID: 627. Target Synonyms: Brain-derived neurotrophic factor; Abrineurin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9164