Learn More
Biomatik Corporation Human Phospholipase A2, Group IIA (PLA2G2A) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06613
Encompass_Preferred
The Human Phospholipase A2, Group IIA (PLA2G2A) ELISA Kit allows for quantitative determination of Phospholipase A2, Group IIA in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 12.5-800 pg/mL. Sensitivity: 5.1 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P14555. Cross Activity: No significant cross-reactivity or interference between Phospholipase A2, Group IIA and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3516