Learn More
Biomatik Corporation Human Pigment Epithelium Derived Factor (PEDF) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06638
Encompass_Preferred
The Human Pigment Epithelium Derived Factor (PEDF) ELISA Kit allows for quantitative determination of Pigment Epithelium Derived Factor in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10μg/mL. Sensitivity: 0.065μg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P36955. Cross Activity: No significant cross-reactivity or interference between Pigment Epithelium Derived Factor and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3521