Learn More
Biomatik Corporation Human Placenta Growth Factor (PLGF) ELISA Kit, 96 tests
Catalog No. 501503525
Encompass_Preferred
The Human Placenta Growth Factor (PLGF) ELISA Kit allows for quantitative determination of Placenta Growth Factor in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 15.6-1000 pg/mL. Sensitivity: 6.5 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P49763. Cross Activity: No significant cross-reactivity or interference between Placenta Growth Factor and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3525 Supplier Biomatik Corporation Supplier No. EKU06645
The Human Placenta Growth Factor (PLGF) ELISA Kit allows for quantitative determination of Placenta Growth Factor in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 15.6-1000 pg/mL. Sensitivity: 6.5 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P49763. Cross Activity: No significant cross-reactivity or interference between Placenta Growth Factor and analogues was observed. Restriction: For Research Use Only.