Learn More
Biomatik Corporation Human Placental Lactogen (PL) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06649
Encompass_Preferred
The Human Placental Lactogen (PL) ELISA Kit allows for quantitative determination of Placental Lactogen in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 2.5-160 ng/mL. Sensitivity: 0.9 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q6PF11. Cross Activity: No significant cross-reactivity or interference between Placental Lactogen and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3527