Learn More
Biomatik Corporation Pig Myoglobin (MYO) ELISA Kit
Supplier:  Biomatik Corporation EKU06080
Encompass_Preferred
Pig Myoglobin (MYO) ELISA Kit for detecting MYO in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 0.78-50ng/ml. Sensitivity: 0.30ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P02189. Gene ID: 397467. Target Synonyms: PVALB; MB. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6244