Learn More
Biomatik Corporation Human Nephronectin (NPNT) ELISA Kit
Supplier:  Biomatik Corporation EKU06166
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 0.156-10ng/mL. Sensitivity: 0.054ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Q6UXI9. Target Synonyms: EGFL6L; POEM; Preosteoblast EGF-like repeat protein with MAM domain; Protein EGFL6-like. Restriction: For Research Use Only.
Catalog No. 50-228-6251