Learn More
Biomatik Corporation Human Plasminogen Activator Inhibitor 2 (PAI2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06662
Encompass_Preferred
The Human Plasminogen Activator Inhibitor 2 (PAI2) ELISA Kit allows for quantitative determination of Plasminogen Activator Inhibitor 2 in Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.78-50 ng/mL. Sensitivity: 0.29 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P05120. Cross Activity: No significant cross-reactivity or interference between Plasminogen Activator Inhibitor 2 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3532