Learn More
Biomatik Corporation Human Plasminogen Activator, Tissue (tPA) ELISA Kit, 96 tests
Catalog No. 501503533
Encompass_Preferred
The Human Plasminogen Activator, Tissue (tPA) ELISA Kit allows for quantitative determination of Plasminogen Activator, Tissue in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.056 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P00750. Cross Activity: No significant cross-reactivity or interference between Plasminogen Activator, Tissue and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3533 Supplier Biomatik Corporation Supplier No. EKU06666
The Human Plasminogen Activator, Tissue (tPA) ELISA Kit allows for quantitative determination of Plasminogen Activator, Tissue in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.056 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P00750. Cross Activity: No significant cross-reactivity or interference between Plasminogen Activator, Tissue and analogues was observed. Restriction: For Research Use Only.