Learn More
Biomatik Corporation Mouse Plasminogen Activator, Tissue (tPA) ELISA Kit, 96 tests
Catalog No. 501503534
Encompass_Preferred
Mouse Tissue Plasminogen Activator (tPA) ELISA Kit for detecting tPA in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.067ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P11214. Gene ID: 18791. Target Synonyms: PLAT; T-PA; t-plasminogen activator; Alteplase; Reteplase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3534 Supplier Biomatik Corporation Supplier No. EKU06667
Mouse Tissue Plasminogen Activator (tPA) ELISA Kit for detecting tPA in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.067ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P11214. Gene ID: 18791. Target Synonyms: PLAT; T-PA; t-plasminogen activator; Alteplase; Reteplase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.