Learn More
Biomatik Corporation Rat Plasminogen Activator, Tissue (tPA) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06669
Encompass_Preferred
Rat Tissue Plasminogen Activator (tPA) ELISA Kit for detecting tPA in plasma. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.115ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P19637. Gene ID: 25692. Target Synonyms: PLAT; T-PA; t-plasminogen activator; Alteplase; Reteplase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3536