Learn More
Biomatik Corporation Rat Plasminogen Activator, Urokinase (uPA) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06674
Encompass_Preferred
Rat Plasminogen Activator, Urokinase (uPA) ELISA Kit for detecting uPA in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.6-1000pg/ml. Sensitivity: 5.8pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P29598. Target Synonyms: PLAU; ATF; URK; UK; UP-A; Abbokinase; Urokinase-Type Plasminogen Activator. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3539