Learn More
Biomatik Corporation Human Platelet Derived Growth Factor BB (PDGFBB) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06685
Encompass_Preferred
The Human Platelet Derived Growth Factor BB (PDGFBB) ELISA Kit allows for quantitative determination of Platelet Derived Growth Factor BB in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 31.2-2000pg/m. Sensitivity: 12.8 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P01127. Cross Activity: No significant cross-reactivity or interference between Platelet Derived Growth Factor BB and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3545