Learn More
Biomatik Corporation Human Perilipin 5 (PLIN5) ELISA Kit
Supplier:  Biomatik Corporation EKU06535
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 0.156-10ng/mL. Sensitivity: 0.055ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Not Available. Target Synonyms: LSDP5; LSDA5; OXPAT; PAT-1; MLDP; Lipid Storage Droplet Protein 5. Restriction: For Research Use Only.
Catalog No. 50-228-6299