Learn More
Biomatik Corporation Mouse Platelet Derived Growth Factor C (PDGFC) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06687
Encompass_Preferred
Mouse Platelet Derived Growth Factor C (PDGFC) ELISA Kit for detecting PDGFC in serum, platelet-poor plasma, cell culture supernates. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.059ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q8CI19. Gene ID: 54635. Target Synonyms: PDGF-C; SCDGF; VEGF-E; Fallotein; Spinal cord-derived growth factor; Platelet-derived growth factor C, latent form; Platelet-derived growth factor C, receptor-binding form. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3546