Learn More
Biomatik Corporation Human Placental Protein 13 (PP13) ELISA Kit
Supplier:  Biomatik Corporation EKU06650
Encompass_Preferred
Human Placental Protein 13 (PP13) ELISA Kit for detecting PP13 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.5pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9UHV8. Gene ID: 29124. Target Synonyms: LGALS13; PLAC8; PP13; GAL13; Galectin 13; Galactoside-Binding Soluble Lectin 13; Placental tissue protein 13. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6312