Learn More
Biomatik Corporation Rabbit Platelet Derived Growth Factor Subunit A (PDGFA) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06698
Encompass_Preferred
Rabbit Platelet Derived Growth Factor Subunit A (PDGFA) ELISA Kit for detecting PDGFA in serum, plasma, other biological fluids. Species Reactivity: Rabbit (Oryctolagus cuniculus). Detection Range: 31.25-2,000pg/ml. Sensitivity: 10.8pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P34007. Target Synonyms: PDGF-A; PDGF1; Platelet Derived Growth Factor Alpha Polypeptide. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3550