Learn More
Biomatik Corporation Rat Platelet Derived Growth Factor Subunit B (PDGFB) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06701
Encompass_Preferred
Rat Platelet Derived Growth Factor Subunit B (PDGFB) ELISA Kit for detecting PDGFB in serum, plasma, tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 31.2-2,000pg/ml. Sensitivity: 12.9pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q05028. Gene ID: 24628. Target Synonyms: PDGF-B; PDGF2; SIS; SSV; C-sis; Simian Sarcoma Viral (v-sis) Oncogene Homolog; Platelet Derived Growth Factor Beta Polypeptide; Proto-oncogene c-Sis; Becaplermin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3551