Learn More
Biomatik Corporation Human Platelet Factor 4 (PF4) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06703
Encompass_Preferred
The Human Platelet Factor 4 (PF4) ELISA Kit allows for quantitative determination of Platelet Factor 4 in Serum, Platelet-poor Plasma and Cell culture supernates. Species Reactivity: Homo sapiens (Human). Detection Range: 1.56-100 pg/mL. Sensitivity: 0.65 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P02776. Cross Activity: No significant cross-reactivity or interference between Platelet Factor 4 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3552