Learn More
Biomatik Corporation Human Proprotein Convertase Subtilisin/Kexin Type 1 (PCSK1) ELISA Kit
Supplier:  Biomatik Corporation EKU06843
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Human. Detection Range: 0.312-20ng/mL. Sensitivity: 0.122ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P29120. Target Synonyms: PC1, PC3, SPC3; NEC1; Prohormone Convertase 3; Prohormone Convertase 1; Neuroendocrine Convertase 1; Proprotein Convertase 1. Restriction: For Research Use Only.
Catalog No. 50-228-6333