Learn More
Biomatik Corporation Mouse Pleiotrophin (PTN) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06717
Encompass_Preferred
Mouse Pleiotrophin (PTN) ELISA Kit for detecting PTN in serum, plasma, tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 7.8-500pg/ml. Sensitivity: 2.9pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P63089. Gene ID: 19242. Target Synonyms: HARP; HBGF8; HBNF; NEGF1; Heparin Binding Growth Factor 8; Neurite Growth-Promoting Factor 1; Heparin Affin Regulatory Peptide; Heparin Binding Growth Associated Molecule. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3558